Learn More
Dräger™ X-act™ 5000 Automatic Tube Pump
Designed for measurements with Draeger short-term tubes and for sampling tubes and systems
£1195.00
Specifications
For Use With (Equipment) | Draeger Short-term Tubes and for Sampling Tubes and Systems |
---|---|
For Use With (Application) | For sampling of toxic gases and vapors in workplace |
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | ||||
13408309
|
Dräger™
4523500 |
X-act 5000 Automatic Tube Pump | Tube pump, shoulder strap |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Simple operation and reduced measurement time
- Swiping bar-code loads measurement data
- Automatic adjustment of flow for technical gases
- No external flow meter needed
- Connection with tube swivel
- With segment and matrix display, barcode reader and flashing lights
X-act 5000 Automatic Tube Pump Kit:
- Includes pump with shoulder strap and rechargeable NiMH battery
- With power supply
Specifications
Draeger Short-term Tubes and for Sampling Tubes and Systems | |
For sampling of toxic gases and vapors in workplace |