Learn More
Dräger™ X-act™ 5000 Automatic Tube Pump
Designed for measurements with Draeger short-term tubes and for sampling tubes and systems
Brand: Dräger™ 4523500
Code : NEW
Additional Details : Weight : 1.89180kg
Description
- Simple operation and reduced measurement time
- Swiping bar-code loads measurement data
- Automatic adjustment of flow for technical gases
- No external flow meter needed
- Connection with tube swivel
- With segment and matrix display, barcode reader and flashing lights
X-act 5000 Automatic Tube Pump Kit:
- Includes pump with shoulder strap and rechargeable NiMH battery
- With power supply
Specifications
Draeger Short-term Tubes and for Sampling Tubes and Systems | |
X-act 5000 Automatic Tube Pump | |
For sampling of toxic gases and vapors in workplace | |
Tube pump, shoulder strap |