missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSTF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CSTF1 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16814754
|
Proteintech
15537-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16804754
|
Proteintech
15537-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.Specifications
CSTF1 | |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q05048 | |
CSTF1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
1477 | |
CSTF1 Fusion Protein Ag7874 | |
Primary | |
-20°C | |
CSTF1 |