Learn More
CSTF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15537-1-AP-20UL
 View more versions of this product
Product Code. 16814754
  • £135.00 / 20µL
Explore more special offers
Description
Description
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5′ UTR, but encoding the same protein, have been found for this gene.
Specifications
Specifications
CSTF1
Polyclonal
Unconjugated
CSTF1
CSTF1
CSTF1 Fusion Protein Ag7874
20 μL
Primary
Human
Antibody
IgG
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q05048
Rabbit
Antigen Affinity Chromatography
RUO
1477
-20°C