missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ BioPharm Plus™ I/P™ Precision Pump Tubing, platinum-cured silicone

Ensure optimal performance from your Masterflex™ pump with this platinum-cured silicone tubing. Masterflex™ BioPharm Plus™ I/P™ Precision Pump Tubing is ideal for pharmaceutical, biotechnology, peristaltic pumping and food applications.

Brand:  Masterflex™ 96440-73

 View more versions of this product

Product Code. 11791019

  • £650.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ideal for pharmaceutical and biotechnological applications
  • Low extractable, excellent heat resistance and low compression set
  • Lot-to-lot consistency for superior accuracy and repeatability
  • Custom extruded to fit Masterflex™ pumps
  • Provides longer life than other platinum and peroxide silicone tubing formulations
  • Manufactured according to Good Manufacturing Practices (GMP)
  • Longer tubing life means less tubing changes and maintenance requirements
  • Complies with USP Class VI, EP, FDA, 3A, REACH, RoHS, ADCF
  • Excellent biocompatibility and low absorption
  • Translucent, odorless and nontoxic
  • Weather, ozone, corona and radiation resistant
  • Can be sterilized by autoclave, gamma radiation or ethylene oxide
Specifications

Specifications

Pump Tubing
9.5 mm
760 cm
-59°C
0.01 to 8L/min.
8.8 m
40 psi
232°C
Autoclavable
BioPharm Plus (Platinum-cured Silicone)
pharmaceutical, biotechnology, peristaltic pumping, and food applications
73
3/8 in.
Translucent
660 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers