Learn More
Cinnamyl chloride, 95% trans, Thermo Scientific Chemicals
£20.60 - £202.00
Chemical Identifiers
CAS | 2687-12-9 |
---|---|
Molecular Formula | C9H9Cl |
Molecular Weight (g/mol) | 152.62 |
MDL Number | MFCD00000986 |
InChI Key | IWTYTFSSTWXZFU-QPJJXVBHSA-N |
Synonym | cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride |
PubChem CID | 639658 |
IUPAC Name | [(E)-3-chloroprop-1-enyl]benzene |
SMILES | C1=CC=C(C=C1)C=CCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10674872
|
Thermo Scientific Acros
181850050 |
5 g | Glass bottle |
£20.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10743571
|
Thermo Scientific Acros
181851000 |
100 g | Glass bottle |
£49.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10245950
|
Thermo Scientific Acros
181855000 |
500 g | Glass bottle |
£202.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2687-12-9 | |
152.62 | |
IWTYTFSSTWXZFU-QPJJXVBHSA-N | |
639658 | |
C1=CC=C(C=C1)C=CCCl |
C9H9Cl | |
MFCD00000986 | |
cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride | |
[(E)-3-chloroprop-1-enyl]benzene |
Specifications
2687-12-9 | |
Brown-Yellow to Yellow | |
120.0°C to 126.0°C (15.0 mmHg) | |
Authentic | |
C6H5CH=CHCH2Cl | |
MFCD00000986 | |
13,299 | |
cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride | |
IWTYTFSSTWXZFU-QPJJXVBHSA-N | |
[(E)-3-chloroprop-1-enyl]benzene | |
639658 | |
95% trans | |
Cinnamyl chloride, 95% trans |
7.0°C | |
1.0960g/mL | |
79°C | |
C9H9Cl | |
1.5830 to 1.5850 | |
05, II, 372 | |
1.096 | |
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, n,n-dmf,, acetone, formic acid, benzene, ethyl ether | |
C1=CC=C(C=C1)C=CCCl | |
152.62 | |
152.62 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Fatal if inhaled.
May cause an allergic skin reaction.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-246-4
- Cinnamyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only