missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamyl chloride, 95% trans, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 7.0°C |
Color | Brown-Yellow to Yellow |
Density | 1.0960g/mL |
Boiling Point | 120.0°C to 126.0°C (15.0 mmHg) |
Flash Point | 79°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | C6H5CH=CHCH2Cl |
Refractive Index | 1.5830 to 1.5850 |
Quantity | 500 g |
Show More |
Safety and Handling
Product Identifier
- Cinnamyl chloride
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'