missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Robu™ Borosilicate Glass Standard Filter Crucibles
Brand: Robu™ 20502
Additional Details : Weight : 0.54150kg
Specifications
46mm | |
Analytical filtration in laboratories | |
46mm | |
Standard Filter Crucible |
Borosilicate Glass | |
50 mL | |
61mm | |
2 |