missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Robu™ Borosilicate Glass Standard Filter Crucibles
Brand: Robu™ 20081
Additional Details : Weight : 0.17950kg
Specifications
24mm | |
Analytical filtration in laboratories | |
24mm | |
Standard Filter Crucible |
Borosilicate Glass | |
8 mL | |
42mm | |
1 |