missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Crude Filter Crucibles

Brand:  Robu™ 20604

Additional Details : Weight : 0.37810kg

 View more versions of this product

Product Code. 16823026

  • £201.00 / Pack of 10
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

34mm
For crude fiber extraction
34mm
Crude Filter Crucible
Borosilicate Glass
30 mL
60mm
4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers