missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGBL5/CCP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | AGBL5/CCP5 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16814804
|
Proteintech
15569-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16804804
|
Proteintech
15569-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGBL5 has a function in the processing of cytosolic proteins such as alpha tubulin, which is known to be modified by the removal of a C terminal tyrosine. It is expressed in the brain. There are three named isoforms.Specifications
AGBL5/CCP5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8NDL9 | |
60509 | |
AGBL5/CCP5 Fusion Protein Ag7889 | |
Primary | |
-20°C | |
AGBL5 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AGBL5, ATP/GTP binding protein like 5, CCP5 | |
AGBL5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |