missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGBL5/CCP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15569-1-AP-20UL
This item is not returnable.
View return policy
Description
AGBL5 has a function in the processing of cytosolic proteins such as alpha tubulin, which is known to be modified by the removal of a C terminal tyrosine. It is expressed in the brain. There are three named isoforms.Specifications
AGBL5/CCP5 | |
Polyclonal | |
Unconjugated | |
AGBL5 | |
AGBL5, ATP/GTP binding protein like 5, CCP5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60509 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NDL9 | |
AGBL5 | |
AGBL5/CCP5 Fusion Protein Ag7889 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |