Learn More
5-Chloropyridine-2-boronic acid, 95%, Thermo Scientific Chemicals
£40.30 - £92.50
Chemical Identifiers
CAS | 652148-91-9 |
---|---|
Molecular Formula | C5H5BClNO2 |
Molecular Weight (g/mol) | 157.36 |
MDL Number | MFCD07368862 |
InChI Key | JEGHCYRKSUGHJH-UHFFFAOYSA-N |
Synonym | 5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx |
PubChem CID | 22832086 |
IUPAC Name | (5-chloropyridin-2-yl)boronic acid |
SMILES | B(C1=NC=C(C=C1)Cl)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15439257
|
Thermo Scientific Alfa Aesar
H52515.MD |
250 mg |
£40.30
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15429257
|
Thermo Scientific Alfa Aesar
H52515.03 |
1 g |
£92.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
652148-91-9 | |
157.36 | |
JEGHCYRKSUGHJH-UHFFFAOYSA-N | |
22832086 | |
B(C1=NC=C(C=C1)Cl)(O)O |
C5H5BClNO2 | |
MFCD07368862 | |
5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx | |
(5-chloropyridin-2-yl)boronic acid |
Specifications
652148-91-9 | |
MFCD07368862 | |
JEGHCYRKSUGHJH-UHFFFAOYSA-N | |
(5-chloropyridin-2-yl)boronic acid | |
22832086 | |
95% |
C5H5BClNO2 | |
5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx | |
B(C1=NC=C(C=C1)Cl)(O)O | |
157.36 | |
157.37 | |
5-Chloropyridine-2-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only