missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyridine-2-boronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15429257
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloropyridine-2-boronic acid was used as reagent in organic synthesis. Also used as phamaceutical Intermediates. Used in suzuki-Miyaura coupling processes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652148-91-9
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD07368862
InChI Key JEGHCYRKSUGHJH-UHFFFAOYSA-N
Synonym 5-chloro-2-pyridineboronic acid, 5-chloropyridine-2-boronic acid, 5-chloropyridin-2-yl boronic acid, 2-borono-5-chloropyridine, 5-chloropyridine-2-boronicacid, 3-chloropyridine-6-boronic acid, 5-chloro-2-pyridyl boronic acid, boronic acid, 5-chloro-2-pyridinyl, pubchem13568, acmc-209npx
PubChem CID 22832086
IUPAC Name (5-chloropyridin-2-yl)boronic acid
SMILES B(C1=NC=C(C=C1)Cl)(O)O

Specifications

Quantity 1 g
Formula Weight 157.37
Percent Purity 95%
Chemical Name or Material 5-Chloropyridine-2-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'