Learn More
4-Benzyloxy-2-fluorobenzeneboronic acid, 96%, Thermo Scientific™
£49.50 - £383.00
Chemical Identifiers
CAS | 166744-78-1 |
---|---|
Molecular Formula | C13H12BFO3 |
Molecular Weight (g/mol) | 246.044 |
MDL Number | MFCD02093064 |
InChI Key | PQWKGFALWCOVTC-UHFFFAOYSA-N |
Synonym | 4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0 |
PubChem CID | 2737786 |
IUPAC Name | (2-fluoro-4-phenylmethoxyphenyl)boronic acid |
SMILES | B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
166744-78-1 | |
246.044 | |
PQWKGFALWCOVTC-UHFFFAOYSA-N | |
2737786 | |
B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O |
C13H12BFO3 | |
MFCD02093064 | |
4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0 | |
(2-fluoro-4-phenylmethoxyphenyl)boronic acid |
Specifications
166744-78-1 | |
MFCD02093064 | |
PQWKGFALWCOVTC-UHFFFAOYSA-N | |
(2-fluoro-4-phenylmethoxyphenyl)boronic acid | |
2737786 | |
96% |
C13H12BFO3 | |
4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0 | |
B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O | |
246.044 | |
246.04 | |
4-Benzyloxy-2-fluorobenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures