missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-butanol, ACS Reagent, ≥98.5%, Honeywell Riedel-de Haën™
ACS Reagent,≥98.5%
£39.88 - £69.69
Identificatori chimici
CAS | 123-51-3 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00002934 |
InChI Key | PHTQWCKDNZKARW-UHFFFAOYSA-N |
Synonym | 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol |
PubChem CID | 31260 |
ChEBI | CHEBI:15837 |
IUPAC Name | 3-methylbutan-1-ol |
SMILES | CC(C)CCO |
Descrizione
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Identificatori chimici
123-51-3 | |
88.15 | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
31260 | |
3-methylbutan-1-ol |
C5H12O | |
MFCD00002934 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CHEBI:15837 | |
CC(C)CCO |
Specifica
123-51-3 | |
0.809g/mL at 25°C | |
1.2 to 9% (100°F) | |
644°F | |
(CH3)2CHCH2CH2OH | |
UN1105 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CC(C)CCO | |
88.15 | |
3 (vs air) | |
88.15g/mol | |
≥98.5% | |
3-Methyl-1-Butanol |
-117°C | |
130°C | |
43°C (109.4°F) | |
C5H12O | |
MFCD00002934 | |
1718835 | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
3-methylbutan-1-ol | |
31260 | |
CHEBI:15837 | |
2mmHg (20°C) | |
ACS Reagent |
Sicurezza e movimentazione
P210-P261-P280-P304 + P340 + P312-P337 + P313-P403 + P235
H226-H315-H319-H332-H335
missing translation for 'dotInformation' : Class 3 - PG 3 - Pentanols
missing translation for 'einecsNumber' : 204-633-5
missing translation for 'rtecsNumber' : EL5425000