missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, ACS Reagent, ≥98.5%, Honeywell Riedel-de Haën™

Product Code. 15663620
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥98.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -117°C
Density 0.809g/mL at 25°C
Boiling Point 130°C
Explosion Limit 1.2 to 9% (100°F)
Flash Point 43°C (109.4°F)
Ignition Point 644°F
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH2OH
Quantity 1 L
UN Number UN1105
Beilstein 1718835
Vapor Density 3 (vs air)
Formula Weight 88.15g/mol
Vapor Pressure 2mmHg (20°C)
Percent Purity ≥98.5%
Grade ACS Reagent
Chemical Name or Material 3-Methyl-1-Butanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'