missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Benzoylpropionic acid, 98%, Thermo Scientific Chemicals
£48.40 - £346.39
Chemical Identifiers
CAS | 2051-95-8 |
---|---|
Molecular Formula | C10H10O3 |
Molecular Weight (g/mol) | 178.19 |
MDL Number | MFCD00002792 |
InChI Key | KMQLIDDEQAJAGJ-UHFFFAOYSA-N |
Synonym | 3-benzoylpropionic acid, 4-oxo-4-phenylbutyric acid, 3-benzoylpropanoic acid, beta-benzoylpropionic acid, benzoylpropionic acid, propanoic acid, 3-benzoyl, benzenebutanoic acid, .gamma.-oxo, 3-benzoylpropionicacid, propionic acid, 3-benzoyl, 4-phenyl-4-oxobutyric acid |
PubChem CID | 72871 |
ChEBI | CHEBI:64437 |
IUPAC Name | 4-oxo-4-phenylbutanoic acid |
SMILES | OC(=O)CCC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10607442
|
Thermo Scientific Acros
|
25 g | Glass bottle |
£48.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10549800
|
Thermo Scientific Acros
105791000 |
100 g | Glass bottle |
£132.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10590024
|
Thermo Scientific Acros
105795000 |
500 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2051-95-8 | |
178.19 | |
KMQLIDDEQAJAGJ-UHFFFAOYSA-N | |
72871 | |
4-oxo-4-phenylbutanoic acid |
C10H10O3 | |
MFCD00002792 | |
3-benzoylpropionic acid, 4-oxo-4-phenylbutyric acid, 3-benzoylpropanoic acid, beta-benzoylpropionic acid, benzoylpropionic acid, propanoic acid, 3-benzoyl, benzenebutanoic acid, .gamma.-oxo, 3-benzoylpropionicacid, propionic acid, 3-benzoyl, 4-phenyl-4-oxobutyric acid | |
CHEBI:64437 | |
OC(=O)CCC(=O)C1=CC=CC=C1 |
Specifications
2051-95-8 | |
100.0 | |
White to Cream | |
97.5% min. (HPLC) | |
C6H5COCH2CH2CO2H | |
10, 696 | |
3-benzoylpropionic acid, 4-oxo-4-phenylbutyric acid, 3-benzoylpropanoic acid, beta-benzoylpropionic acid, benzoylpropionic acid, propanoic acid, 3-benzoyl, benzenebutanoic acid, .gamma.-oxo, 3-benzoylpropionicacid, propionic acid, 3-benzoyl, 4-phenyl-4-oxobutyric acid | |
OC(=O)CCC(=O)C1=CC=CC=C1 | |
178.19 | |
CHEBI:64437 | |
98% | |
3-Benzoylpropionic acid |
97.5 | |
114°C to 119°C | |
Authentic | |
C10H10O3 | |
MFCD00002792 | |
02,26; 03,19 | |
KMQLIDDEQAJAGJ-UHFFFAOYSA-N | |
4-oxo-4-phenylbutanoic acid | |
72871 | |
178.19 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 218-135-
RUO – Research Use Only