missing translation for 'onlineSavingsMsg'
Learn More

3-Benzoylpropionic acid, 98%, Thermo Scientific Chemicals

Product Code. 10549800
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-95-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00002792
InChI Key KMQLIDDEQAJAGJ-UHFFFAOYSA-N
Synonym 3-benzoylpropionic acid, 4-oxo-4-phenylbutyric acid, 3-benzoylpropanoic acid, beta-benzoylpropionic acid, benzoylpropionic acid, propanoic acid, 3-benzoyl, benzenebutanoic acid, .gamma.-oxo, 3-benzoylpropionicacid, propionic acid, 3-benzoyl, 4-phenyl-4-oxobutyric acid
PubChem CID 72871
ChEBI CHEBI:64437
IUPAC Name 4-oxo-4-phenylbutanoic acid
SMILES OC(=O)CCC(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 114°C to 119°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5COCH2CH2CO2H
Quantity 100 g
Beilstein 10, 696
Fieser 02,26; 03,19
Formula Weight 178.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzoylpropionic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'