Learn More
2-Chloro-N,N-dimethyl-4-nitroaniline, 96%, Thermo Scientific™
£17.60 - £184.00
Chemical Identifiers
CAS | 6213-19-0 |
---|---|
Molecular Formula | C8H9ClN2O2 |
Molecular Weight (g/mol) | 200.622 |
MDL Number | MFCD00043604 |
InChI Key | OZKAWTHGBGLZKC-UHFFFAOYSA-N |
Synonym | 2-chloro-nn-dimethyl-4-nitroaniline, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline, 2-chloro-4-nitrophenyl dimethylamine, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, benzenamine,2-chloro-n,n-dimethyl-4-nitro, 2-chloro-4-nitro-n,n-dimethylaniline |
PubChem CID | 80343 |
IUPAC Name | 2-chloro-N,N-dimethyl-4-nitroaniline |
SMILES | CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6213-19-0 | |
200.622 | |
OZKAWTHGBGLZKC-UHFFFAOYSA-N | |
80343 | |
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl |
C8H9ClN2O2 | |
MFCD00043604 | |
2-chloro-nn-dimethyl-4-nitroaniline, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline, 2-chloro-4-nitrophenyl dimethylamine, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, benzenamine,2-chloro-n,n-dimethyl-4-nitro, 2-chloro-4-nitro-n,n-dimethylaniline | |
2-chloro-N,N-dimethyl-4-nitroaniline |
Specifications
6213-19-0 | |
MFCD00043604 | |
2-chloro-nn-dimethyl-4-nitroaniline, benzenamine, 2-chloro-n,n-dimethyl-4-nitro, n,n-dimethyl-4-nitro-2-chloroaniline, 2-chloro-4-nitrophenyl dimethylamine, 3-chloro-4-dimethylamino nitrobenzene, 3-chloro-4-dimethylaminonitrobenzene, 2-chloro-n,n-dimethyl-4-nitrobenzenamine, benzenamine,2-chloro-n,n-dimethyl-4-nitro, 2-chloro-4-nitro-n,n-dimethylaniline | |
CN(C)C1=C(C=C(C=C1)[N+](=O)[O-])Cl | |
200.622 | |
200.62 | |
Solid |
C8H9ClN2O2 | |
UN2811 | |
OZKAWTHGBGLZKC-UHFFFAOYSA-N | |
2-chloro-N,N-dimethyl-4-nitroaniline | |
80343 | |
96% | |
2-Chloro-N,N-dimethyl-4-nitroaniline |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 228-280-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures