missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haen
CHROMASOLV™, for HPLC,≥99.7%
£54.23 - £607.34
Chemical Identifiers
CAS | 78-93-3 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00011648 |
InChI Key | ZWEHNKRNPOVVGH-UHFFFAOYSA-N |
PubChem CID | 6569 |
ChEBI | CHEBI:28398 |
IUPAC Name | butan-2-one |
SMILES | CCC(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15654580
|
Honeywell-Chromasolv
34861-100ML |
100 mL | Glass Bottle |
£54.23
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15664580
|
Honeywell-Chromasolv
34861-1L |
1 L | Glass Bottle |
£122.88
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15674580
|
Honeywell-Chromasolv
34861-2.5L |
2.5 L | Glass Bottle |
£181.52
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15694580
|
Honeywell Riedel-de Haën
34861-6X1L |
6 x 1 L | Glass Bottle |
£594.76
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15684580
|
Honeywell-Chromasolv
34861-4X2.5L |
4 x 2.5 L | Glass Bottle |
£607.34
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
78-93-3 | |
72.11 | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
CHEBI:28398 | |
CCC(C)=O |
C4H8O | |
MFCD00011648 | |
6569 | |
butan-2-one |
Specifications
-87°C (literature) | |
100.0000% | |
79°C to 81°C | |
C4H8O | |
MFCD00011648 | |
741880 | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
butan-2-one | |
6569 | |
72.11 | |
HPLC |
78-93-3 | |
0.805 g/mL at 25 °C(lit.) | |
−4°C | |
C2H5COCH3 | |
UN1193 | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
CCC(C)=O | |
72.11 | |
CHEBI:28398 | |
>99.7% | |
2-Butanone, CHROMASOLV™, for HPLC, ≥99.7% |
Safety and Handling
missing translation for 'shelfLife' : 1260 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone
missing translation for 'einecsNumber' : 201-159-0
Recommended Storage : Room Temp