missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haen

Product Code. 15674580
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle

CHROMASOLV™, for HPLC,≥99.7%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87°C (literature)
CAS Max % 100.0000%
Density 0.805 g/mL at 25 °C(lit.)
Boiling Point 79°C to 81°C
Quantity 2.5 L
Flash Point −4°C
Packaging Glass Bottle
Linear Formula C2H5COCH3
UN Number UN1193
Beilstein 741880
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
Formula Weight 72.11
Percent Purity >99.7%
Grade HPLC
Chemical Name or Material 2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Butanone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : butanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'