missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF320 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24882-1-AP-150UL
This item is not returnable.
View return policy
Description
ZNF320 encodes a Kruppel-like zinc finger protein. Members of this protein family are involved in activation or repression of transcription.Specifications
ZNF320 | |
Polyclonal | |
Unconjugated | |
ZNF320 | |
ZFPL, zinc finger protein 320, ZNF320 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
162967 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2RRD8 | |
ZNF320 | |
ZNF320 Fusion Protein Ag21688 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |