Learn More
Pro-Lab Diagnostics™ ZN Carbol Fuchsin Stain Concentrate
Brand: Pro-Lab Diagnostics™ PL8005
Code : 04
Additional Details : Weight : 0.10000kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II
Specifications
100 mL | |
CE marked (98/78/EC) | |
Dark Red | |
Keep at temperature not exceeding 20°C |
Carbol Fuchsin | |
Liquid | |
Phenol 50 - 100% , Ethanol 25 - <50% , Basic Fuchsin 10 - <25% , Methanol 2.5 - <5% | |
Acid Fast |