missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals

Product Code. 11384939
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Zirconium(IV) fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production. Together with uranium salt, zirconium fluoride can be a component of fuel-coolant in molten salt reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-64-4
Molecular Formula F4Zr
MDL Number MFCD00011307
Synonym Zirconium tetrafluoride

Specifications

Melting Point 600°C (sublimation)
Color White
Physical Form Powder
Quantity 250 g
Linear Formula ZrF4
UN Number UN3260
Merck Index 14,10175
Solubility Information Slightly soluble in water; Soluble in hydrofluoric acid
Formula Weight 167.21
Percent Purity 98%
Refractive Index 1.59
Sensitivity Moisture sensitive
Density 4.43 g/mL
Chemical Name or Material Zirconium (IV) fluoride
Show More Show Less

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado