missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Monohydrate, ≥99.9% trace metals basis, Honeywell™

≥99.9% trace metals basis

£42.94 - £171.76

Chemical Identifiers

CAS 7446-19-7
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.451
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
IUPAC Name zinc;sulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[Zn+2]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15683120
View Documents
Honeywell Research Chemicals
307491-100G
100 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15693120
View Documents
Honeywell Research Chemicals
307491-500G
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

7446-19-7
179.451
RNZCSKGULNFAMC-UHFFFAOYSA-L
62639
O.[O-]S(=O)(=O)[O-].[Zn+2]
H2O5SZn
MFCD00149893
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
zinc;sulfate;hydrate

Specifications

238°C
H2O5SZn
MFCD00149893
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
RNZCSKGULNFAMC-UHFFFAOYSA-L
zinc;sulfate;hydrate
62639
≥99.9% (trace metals basis)
7446-19-7
ZnSO4 · H2O
UN3077
Soluble in water
O.[O-]S(=O)(=O)[O-].[Zn+2]
179.451
179.47g/mol
Zinc Sulfate Monohydrate
Videos
Safety and Handling

Safety and Handling

P273-P280-P305 + P351 + P338-P501

H302-H318-H410

missing translation for 'dotInformation' : 9/PGIII

missing translation for 'einecsNumber' : 231-793-3

SDS
Documents

Documents

Product Certifications