missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

£50.35 - £132.66

Specifications

Chemical Name or Material Zinc chloride
Color Blue-Gray or Colorless to Yellow
Name Note 1.0M solution in diethyl ether
CAS 7646-85-7
Molecular Formula Cl2Zn
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10292572
View Documents Promotion Details
Thermo Scientific Acros
370051000
100 mL AcroSeal™ Glass Bottle
£50.35
100mL
Please to purchase this item. Need a web account? Register with us today!
10029051
View Documents
Thermo Scientific Acros
370058000
800 mL AcroSeal™ Glass Bottle
£132.66
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc chloride
1.0M solution in diethyl ether
Cl2Zn
MFCD00011295
01,1289; 02,464; 03,338; 05,763; 06,676; 08,536; 09,522; 10,461; 11,602; 12,446; 13,44; 14,177; 15,89; 16,391
15, 10331
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
Solution, May Contain A Small Amount Precipitate
Blue-Gray or Colorless to Yellow
7646-85-7
ZnCl2
0.9 to 1.1M
0.8400g/mL
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
Cl[Zn]Cl
136.28
CHEBI:49976
AcroSeal™ Glass Bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Very toxic to aquatic life with long lasting effects. Extremely flammable liquid and vapor. May cause respiratory irritation. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge. Store in a well-ventilated place. Keep cool.

Danger

Recommended Storage : Product can form two phases, which can recombine on cooling

Product Identifier
  • Zinc chloride, 1.0M solution in diethyl ether

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only