Learn More
Zinc acetate dihydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
£39.20 - £316.40
Chemical Identifiers
CAS | 5970-45-6 |
---|---|
Molecular Formula | C4H6O4Zn·2H2O |
Molecular Weight (g/mol) | 219.5 |
MDL Number | MFCD00066961 |
InChI Key | BEAZKUGSCHFXIQ-UHFFFAOYSA-L |
Synonym | zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn |
PubChem CID | 2724192 |
IUPAC Name | zinc;diacetate;dihydrate |
SMILES | CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10369460
|
Thermo Scientific Acros
317215000 |
500 g | Plastic bottle |
£39.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10688824
|
Thermo Scientific Acros
317210025 |
2.5 kg | Plastic bottle |
£123.62
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15109273
|
Thermo Scientific Acros
317210100 |
10 kg | Plastic drum | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5970-45-6 | |
219.5 | |
BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
2724192 | |
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] |
C4H6O4Zn·2H2O | |
MFCD00066961 | |
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn | |
zinc;diacetate;dihydrate |
Specifications
237.0°C | |
White | |
Crystals | |
C4H6O4Zn·2H2O | |
MFCD00066961 | |
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn | |
BEAZKUGSCHFXIQ-UHFFFAOYSA-L | |
zinc;diacetate;dihydrate | |
2724192 | |
≥98% | |
1.84 | |
Zinc acetate dihydrate, 98+% |
5970-45-6 | |
6.0 to 7.0 (5% soln. at 25°C) | |
98.0 to 101% (Complexometry) | |
(CH3CO2)2Zn·2H2O | |
15,10327 | |
Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol | |
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] | |
219.5 | |
219.5 | |
ACS Reagent | |
1.8400g/mL |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Very toxic to aquatic life.
GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
missing translation for 'rtecsNumber' : ZG8750000
- Zinc acetate dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.