missing translation for 'onlineSavingsMsg'
Learn More

ZFYVE21 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ZFYVE21
Concentration 0.4 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883305
View Documents
Proteintech
24806-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16873305
View Documents
Proteintech
24806-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZF21 was initially identified as protein that could bind to the cytoplasmic tail of MT1-MMP (Membrane-type 1 matrix metalloproteinase), a potent invasion-promoting protease. ZF21 is a member of a protein family characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain and regulates focal adhesions (FAs) and cell movement. Knockdown of ZF21 expression resulted in a delay of FA disassembly following induction of synchronous disassembly of FAs by nocodazole treatment, suggesting that ZF21 is involved in FA disassembly. ZF21 contains a noncanonical pleckstrin homology domain that is a possible therapeutic target to treat metastatic cancer.
Specifications

Specifications

ZFYVE21
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Human
Q9BQ24
79038
ZFYVE21 Fusion Protein Ag20024
Primary
-20°C
ZFYVE21
0.4 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ZFYVE21
ZFYVE21
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.