Learn More
ZCCHC7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554242
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SNRWPQENKE TQKEMKNKNR NWEKHRKADR HREVDEDFPR GPKTYSSPGS FKTQKPSKPF HRSSHYHTSR EDKSPKEGKR GKQKKKERCW EDDDNDNLFL IKQ Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 53%.
ZCCHC7 Polyclonal antibody specifically detects ZCCHC7 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
ZCCHC7 | |
Polyclonal | |
Unconjugated | |
ZCCHC7 | |
4930572I07Rik; AIR1; D4Wsu132e; HSPC086; TRAMP-like complex RNA-binding factor ZCCHC7; ZCCHC7; Zinc finger CCHC domain-containing protein 7; zinc finger CCHC-type containing 7; zinc finger, CCHC domain containing 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84186 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N3Z6 | |
ZCCHC7 | |
Recombinant protein corresponding to Human ZCCHC7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only