missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB44 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25421-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in transcriptional regulation.Specifications
ZBTB44 | |
Polyclonal | |
Unconjugated | |
ZBTB44 | |
BTBD15, HSPC063, ZBTB44, ZNF851 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29068 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NCP5 | |
ZBTB44 | |
ZBTB44 Fusion Protein Ag22299 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |