man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
pH and Electrochemistry
Probes and Electrodes
Dissolved Oxygen Probe Accessories
YSI™ Membrane kits
Print
YSI™ Membrane kits
£60.15
See promotional offers below
Specifications
For Use With (Equipment)
YSI™ Model 556
Type
Membrane Kit
View More Specs
Products
1
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
1
Product Code
Brand
Probe Type
Includes
Price
Quantity & Availability
Product Code
Brand
Probe Type
Includes
Price
Quantity & Availability
12774516
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
YSI™
605306
DO
Six 1.25 mil. cap membranes, KCl solution
£60.15
Pack of 6
View alternative products
Save Up to
»
Estimated Shipment: 13-06-2024
Log in
to see stock.
Add to basket
Description
Description
Specifications
Specifications
For Use With (Equipment)
YSI™ Model 556
Type
Membrane Kit
Videos
SDS
Documents
Documents
Product Certifications
Back to Top