missing translation for 'onlineSavingsMsg'
Learn More

Manutan Yellow/black barrier tape, 500m x 75mm

Brand:  Manutan A353880

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12858978

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Yellow/black
  • Dimensions (L x W) : 500m x 75mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers