missing translation for 'onlineSavingsMsg'
Learn More

XBP1S-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen XBP1S-Specific
Concentration 293 μg/mL
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883365
View Documents
Proteintech
24868-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16873365
View Documents
Proteintech
24868-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The XBP1S-specific antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human XBP1S-specific. This antibody recognizes human, mouse antigen. The XBP1S-specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

XBP1S-Specific
Western Blot, Immunohistochemistry
Unconjugated
Human, Mouse
IgG
293 μg/mL
Polyclonal
Rabbit
7494
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.