missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WISP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551169
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total WISP3 protein.
WISP3 Polyclonal antibody specifically detects WISP3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
WISP3 | |
Polyclonal | |
Unconjugated | |
CCN6 | |
CCN family member 6; CCN6; Cellular communication network factor 6; ENSMUSG00000062074; Gm735; LIBC; PPAC; PPD; RGD1564120; UNQ462/PRO790/PRO956; Wisp3; WISP-3; WNT1 inducible signaling pathway protein 3; WNT1-inducible-signaling pathway protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8838 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95389 | |
CCN6 | |
Fusion protein of human WISP3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |