missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20512-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.Specifications
WIPF2 | |
Polyclonal | |
Unconjugated | |
WIPF2 | |
WICH, WIP related protein, WIPF2, WIRE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
147179, 360620, 68524 | |
-20°C | |
Liquid |
Western Blot | |
0.73 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6PEV3, Q8TF74 | |
Wipf2 | |
WIPF2 Fusion Protein Ag14330 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |