missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Plastic, White/Blue Self-Adhesive Graduation
6.73 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Use promo code "23267" to get your promotional price.
Click to view available options
Color: White and Blue
Description
- Self-adhesive
- White and blue
- For use with wide-necked jerrycans
Specifications
Material | Plastic |
Type | Graduation |
No. per Pack | 1 |
For Use With (Equipment) | 20 L Wide Necked Jerry Cans |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'