missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Screw Caps with bonded PTFE Faced Silicone Liner for E-Z Ex-Traction™ Vials

Product Code. 10212463
Click to view available options
Neck Finish:
13-425
15-425
22-400
24-400
No. per Case:
100 Cs.
1000 Cs.
250 Cs.
500 Cs.
No. per Pack:
100/Cs.
1000/Cs.
250/Cs.
500/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

PTFE faced silicone liner ideal for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 500 Cs.
No. per Pack 500/Cs.
Neck Finish 22-400
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
For Use With (Equipment) 10 mL Vial
Closure Liner PTFE/Silicone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'