missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Plastic Uni-Dose Vials

Safer alternative to glass and cost less to ship

Brand:  DWK Life Sciences 226773

Product Code. 11581225

  • £458.00 / Pack of 300
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Mouth OD 28.5mm
  • Use with WHEATON tamper evident safety cap
  • Large labeling surface for easy product identification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amber
Polyethylene
30.0 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers