missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ E-Z Vial™ with Step

Available wth or without writing patch

Brand:  DWK Life Sciences 224626

 View more versions of this product

Product Code. 12402711

  • £269.00 / Pack of 1000
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Wheaton Chromatography Vials and accessories are designed for trouble-free operation in most autosamplers. The innovative design of the E-Z Vial and limited volume insert enables the insert to be precisely centered inside the vial.

  • Size: 1.8mL, 12 dia. x 32mm H
  • Manufactured from clear or amber Type I borosilicate glass
  • For use in both GC and HPLC applications
  • Limited volume inserts with step available separately

Order Info

Vials and caps are sold separately.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Borosilicate Glass
Both GC and HPLC applications
1000
32 mm
10-425
12 mm
1.8 mL
1.25 in.
Screw Cap
1000 Cs.
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers