missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Wedge Shaped Spreaders

Wedge shaped to reduce tendency to flip

Brand:  Microspec™ D7

Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 15672767

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Reduces tendency to flip
Specifications

Specifications

Yes
37 x 180 mm
Spreader
Microseal Pouch
High-impact Polystyrene
7.1 in.
Aseptically Manufactured
Blue
Individual Pack
180 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers