missing translation for 'onlineSavingsMsg'
Learn More

WDR52 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA564321

Product Code. 15772082

  • £447.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: HSFGYDCRKR ANLQLLDDSI AIYIAGNQLI FLNLKTKEQI YLRSSSGEGI GVIGVHPHKT YFTVAEKGSF PDIIIYEYPS LRPYRVLRDG TEKG Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.

Flagellar protein involved in sperm flagellum axoneme organization and function. [UniProt]
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WDR52
Polyclonal
Unconjugated
CFAP44
CFAP44; cilia and flagella associated protein 44; cilia- and flagella-associated protein 44; WD repeat domain 52; WD repeat-containing protein 52; WDR52
Rabbit
Antigen affinity chromatography
RUO
55779
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96MT7
CFAP44
Recombinant protein corresponding to Human WDR52
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only