missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTCN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12080-1-AP-150UL
This item is not returnable.
View return policy
Description
B7-H4 (B7x, VTCN1) a costimulatory protein which is reported to function as a negative regulator of T-cell mediated immunity. Although B7-H4 binds an unknown receptor, it is thought to deliver an inhibitory signal to T-cells preventing their proliferation, cell cycle progression and interleukin-2 production. B7-H4 deficient mice are only minimally affected; suggesting B7-H4 is important in the fine tuning of the T-cell mediated immune response. B7-H4 is expressed on activated T-cells, B-cells, monocytes and dendritic cells. Aberrant expression has been associated with cancers of the lung, breast and ovary in humans. B7-H4 is expressed primarily on the membrane of lymphoid cells, and as an immunoinhibitory protein that interacts with receptors on the surface of T lymphocytes, it is involved in mediating cellular and humoral immune responses. Overexpression of B7-H4 is associated with certain maligcies, including ovarian and breast cancer, which may be a mechanism by which tumor cells suppress T cell immunity and facilitate tumor progression. Further, VTCN1 may be a useful biomarker for the early detection of ovarian cancer and Renal Cell Carcinoma (RCC).Specifications
VTCN1 | |
Polyclonal | |
Unconjugated | |
VTCN1 | |
B7 H4, B7h.5, B7H4, B7S1, B7X, PRO1291, Protein B7S1, RP11 229A19.4, VCTN1, VTCN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
242122, 295322, 79679 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q501W4, Q7TSP5, Q7Z7D3 | |
VTCN1 | |
VTCN1 Fusion Protein Ag2712 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |