missing translation for 'onlineSavingsMsg'
Learn More
Vinyltrimethylsilane, 98+%, Thermo Scientific Chemicals Product Code.: 11353886

Vinyltrimethylsilane, 98+%, Thermo Scientific Chemicals

Product Code. 11353886
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11353886

Brand: Thermo Scientific Alfa Aesar L16989.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-05-2
Molecular Formula C5H12Si
Molecular Weight (g/mol) 100.236
MDL Number MFCD00008606
InChI Key GCSJLQSCSDMKTP-UHFFFAOYSA-N
Synonym vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
PubChem CID 79102
IUPAC Name ethenyl(trimethyl)silane
SMILES C[Si](C)(C)C=C

Specifications

Boiling Point 54°C to 56°C
Melting Point -132°C
Quantity 25 g
UN Number UN1993
Beilstein 956572
Flash Point −24°C (−11°F)
Formula Weight 100.24
Percent Purity ≥98%
Odor Mild
Refractive Index 1.391
Density 0.69
Chemical Name or Material Vinyltrimethylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vinyltrimethylsilane, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.