Learn More
Vinyltrimethoxysilane, 98%, AcroSeal™, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 2768-02-7 |
---|---|
Molecular Formula | C5H12O3Si |
Molecular Weight (g/mol) | 148.24 |
MDL Number | MFCD00008605 |
InChI Key | NKSJNEHGWDZZQF-UHFFFAOYSA-N |
Synonym | vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m |
PubChem CID | 76004 |
IUPAC Name | ethenyl(trimethoxy)silane |
SMILES | CO[Si](C=C)(OC)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10713922
|
Thermo Scientific Acros
433061000 |
100 mL | AcroSeal™ Glass Bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10423350
|
Thermo Scientific Acros
433060010 |
1 L | AcroSeal™ Glass Bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2768-02-7 | |
148.24 | |
NKSJNEHGWDZZQF-UHFFFAOYSA-N | |
76004 | |
CO[Si](C=C)(OC)OC |
C5H12O3Si | |
MFCD00008605 | |
vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m | |
ethenyl(trimethoxy)silane |
Specifications
2768-02-7 | |
0.9680g/mL | |
23°C | |
97.5% min. (GC) | |
H2C=CHSi(OCH3)3 | |
MFCD00008605 | |
vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m | |
NKSJNEHGWDZZQF-UHFFFAOYSA-N | |
ethenyl(trimethoxy)silane | |
76004 | |
148.24 | |
Liquid |
Colorless | |
123.0°C | |
Authentic | |
C5H12O3Si | |
1.3915 to 1.3935 | |
0.968 | |
Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents | |
CO[Si](C=C)(OC)OC | |
148.24 | |
0.6 mm2/s (25°C) | |
98% | |
Vinyltrimethoxysilane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 220-449-8
- Vinyltrimethoxysilane
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
- Skin sensitiser Category 1
Hazard Statement
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.