missing translation for 'onlineSavingsMsg'
Learn More
Vinyltrichlorosilane, 97%, Thermo Scientific Chemicals Product Code.: 11346926

Vinyltrichlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11346926
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11346926

Brand: Thermo Scientific Alfa Aesar L04387.22

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-94-5
Molecular Formula C2H3Cl3Si
Molecular Weight (g/mol) 161.481
MDL Number MFCD00000482
InChI Key GQIUQDDJKHLHTB-UHFFFAOYSA-N
Synonym trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichlorideShow MoreShow Less
PubChem CID 6413
IUPAC Name trichloro(ethenyl)silane
SMILES C=C[Si](Cl)(Cl)Cl

Specifications

Boiling Point 91°C to 93°C
Melting Point -95°C
Quantity 100 g
Linear Formula CH2=CHSiCl3
UN Number UN1305
Beilstein 1743440
Flash Point 10°C (50°F)
Formula Weight 161.49
Percent Purity 97%
Refractive Index 1.436
Sensitivity Moisture sensitive
Density 1.27
Chemical Name or Material Vinyltrichlorosilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10162206 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04387.36,L04387.22
Lot Number10163390 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04387.36,L04387.22
Lot NumberF12Z053 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04387.36,L04387.22
Lot Number10172910 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04387.36,L04387.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vinyltrichlorosilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.