missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Mouse anti-Human, Mouse, Rat, Clone: 3H9D1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | Vimentin |
---|---|
Clone | 3H9D1 |
Concentration | 0.8 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16807435
|
Proteintech
60330-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16897425
|
Proteintech
60330-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.Specifications
Vimentin | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
VIM, Vimentin | |
VIM | |
IgG1 | |
Protein G | |
Antibody |
3H9D1 | |
Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Mouse, Human, Rat | |
P08670, P20152, P31000 | |
22352, 7431, 81818 | |
Vimentin Fusion Protein Ag0489 | |
Primary | |
-20°C | |
VIM |