missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Door with Viewing Window and Interior Lighting

Material: Triple Insulating Glass; For Use With: FED 720

Brand:  BINDER™ 8012-1832

Product Code. 16464459

  • £2065.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Viewing window in each door
  • 15 W or 30 W interior lighting
Specifications

Specifications

Viewing window and interior lighting - 208/400 V option model, viewing window (triple insulating glass) in door, 300 x 480 mm, and 30 W interior
Triple Insulating Glass
300 x 480 mm
30 W
BINDER Chambers FED 720
Door with Window and Lighting
208 to 400 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers