Learn More
Cytiva (formerly Pall Lab) Versapor™ Acrylic Copolymer 3000 Membrane Disc Filter
Disc Filters useful for prefiltration of particle-laden samples, serum filtration, and dissolution testing.
Brand:  Cytiva (formerly Pall Lab) 66387
Additional Details : Weight : 0.05100kg
 View more versions of this product
Product Code. 17114561
  • £167.00 / Pack of 100
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Protects instrumentation against particulate build up
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Versapor (Acrylic Copolymer on a Non-woven Support)
Non-sterile
47 mm
3 μm
94 μm