Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Filter
3M Versaflo™ TR-600 PAPR Accessory, Filter is used with 3M™ Versaflo™ TR-600 series air purifying device.
Brand: 3M™ 7100020997
69.80 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : NEW
Additional Details : Weight : 3.60000kg
Description
- Simple to install
- For use with 3M Versaflo Powered Air TR-600 series
- Recommends use of TR-6100FC filter cover to help protect filter from physical damage
Specifications
ABE1P Filter | |
3M Versaflo Powered Air Turbo TR-600 Series | |
TR-6130E |